Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.

$242.22 - $874.18

Specifications

Antigen Amyloid A Component Ab-1
Clone mcl
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1219R7

 
thermo scientific
MS1219R7
7mL Each for $508.31

MS1219S

 
thermo scientific™ lab vision™
MS1219S
1mL Each for $874.18

MS1219S0

 
thermo scientific™ lab vision™
MS1219S0
100μL Each for $242.22

MS1219S1

 
thermo scientific™ lab vision™
MS1219S1
500μL Each for $665.69
Description & Specifications

Specifications

Antigen Amyloid A Component Ab-1
Clone mcl
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Neuroscience
Type Primary
Immunogen Human amyloid A
Isotype IgG2a kappa

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Amyloid A Component Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: mcl

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Immunogen: Human amyloid A

Positive Control: Amyloid deposits in kidney

Cellular Localization: Extracellular

Ab-1 reacts with isolated protein AA in both native fibril form and denatured form. It reacts with amyloid deposits in all organs and tissues including kidney and rectum, and as well as the serum precursor of protein AA.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO