Please login to your online account to display your discounted pricing

Alfa Aesar™ Amylin, Human, Amide


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66825MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66825MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1mg
Storage Requirements -30° to -10°C
Product Type Amylin, human, amide
Regulatory Status RUO
Cross Reactivity Human
Purification Method HPLC
Format Solid
Molecular Weight 3860.16