Please login to your online account to display your discounted pricing

Alfa Aesar™ Amylin (1-13), Human


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66863EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66863EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 0.5mg
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Molecular Weight 1378.57
Cross Reactivity Human
Sequence Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala (Disulfide bridge Cys2-Cys7)
Product Type Amylin (1-13), human
Purification Method HPLC