missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amyl Acetate (Laboratory), Fisher Chemical™

$174.69 - $551.77

Chemical Identifiers

CAS 628-63-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009500
InChI Key PGMYKACGEOXYJE-UHFFFAOYSA-N
Synonym 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate
PubChem CID 12348
ChEBI CHEBI:87362
IUPAC Name pentyl acetate
SMILES CCCCCOC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A718-500
SDS Accessories View Documents
Fisher Chemical
BPA718500
500mL Glass Bottle Each for $174.69
Case of 6 Each for $1,048.14
 
A718-4
SDS Accessories View Documents
Fisher Chemical
BPA7184
4L Poly Bottle Each for $551.77
Case of 4 Each for $2,207.08
 
Specifications

Chemical Identifiers

628-63-7
130.187
PGMYKACGEOXYJE-UHFFFAOYSA-N
12348
pentyl acetate
C7H14O2
MFCD00009500
1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate
CHEBI:87362
CCCCCOC(=O)C

Specifications

0.870g/cm³
Pass Test
149°C
-70.8°C
628-63-7
CH3(CH2)4OC(O)CH3
1104
PGMYKACGEOXYJE-UHFFFAOYSA-N
pentyl acetate
12348
130.19
Laboratory
40°C
Glass Bottle
Colorless
500mL
C7H14O2
MFCD00009500
1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate
CCCCCOC(=O)C
130.187
CHEBI:87362
Liquid
Amyl Acetate
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid