Please login to your online account to display your discounted pricing

Amthamine dihydrobromide, Tocris


Highly selective standard H2 agonist

Quantity 10mg

Manufacturer: r & d systems  RDS066810

 View more versions of this product

Catalog No. 066810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 319.06
Chemical Name or Material Amthamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3S·2HBr
CAS 142457-00-9
Synonym 2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Amthamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3S·2HBr
Formula Weight 319.06
Synonym 2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
Accessories

Desiccate at -20°C