Please login to your online account to display your discounted pricing

Amthamine dihydrobromide, Tocris

Highly selective standard H2 agonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material Amthamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3S·2HBr
Formula Weight 319.06
Synonym 2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

066810

 
r & d systems
RDS066810
10mg Each for $181.19

066850

 
r & d systems
RDS066850
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 319.06
Chemical Name or Material Amthamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3S·2HBr
CAS 142457-00-9
Synonym 2-Amino-5-(2-aminoethyl)-4-methylthiazole dihydrobromide
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C