missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ AMPSO, 98+%

Useful as a buffer

$60.28 - $195.85

Chemical Identifiers

CAS 68399-79-1
Molecular Formula C7H17NO5S
Molecular Weight (g/mol) 227.275
MDL Number MFCD00041777
InChI Key ACERFIHBIWMFOR-UHFFFAOYSA-N
Synonym N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
PubChem CID 100211
IUPAC Name 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
SMILES CC(C)(CO)NCC(CS(=O)(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6237814 View Documents Alfa Aesar™
J6237814
25g Each for $60.28
 
AAJ6237822 View Documents Alfa Aesar™
J6237822
100g Each for $195.85
 
Description

Description

Applications
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.

Solubility
Soluble in water (333.33 mg/ml, clear to slightly hazy colorless solution).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

68399-79-1
227.275
ACERFIHBIWMFOR-UHFFFAOYSA-N
100211
CC(C)(CO)NCC(CS(=O)(=O)O)O
C7H17NO5S
MFCD00041777
N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid

Specifications

68399-79-1
MFCD00041777
N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid
CC(C)(CO)NCC(CS(=O)(=O)O)O
227.275
227.28
≥98%
White
AMPSO
C7H17NO5S
5929916
ACERFIHBIWMFOR-UHFFFAOYSA-N
2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
100211
Crystals
215°C to 225°C
25g
Safety and Handling

Safety and Handling

TSCA : TSCA