missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ AMPSO, 0.2M buffer soln., pH 8.5

$58.46 - $91.61

Chemical Identifiers

CAS 102029-60-7
Molecular Formula C7H16NNaO5S
Molecular Weight (g/mol) 249.257
MDL Number MFCD00067500
InChI Key UVGYCHPTWHWEPZ-UHFFFAOYSA-M
PubChem CID 44119996
IUPAC Name sodium;2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
SMILES CC(C)(CO)NCC(CS(=O)(=O)[O-])O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61238AE View Documents Alfa Aesar™
J61238AE
100mL Each for $58.46
 
AAJ61238AK View Documents Alfa Aesar™
J61238AK
250mL Each for $91.61
 
Specifications

Chemical Identifiers

102029-60-7
249.257
UVGYCHPTWHWEPZ-UHFFFAOYSA-M
sodium;2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
C7H16NNaO5S
MFCD00067500
44119996
CC(C)(CO)NCC(CS(=O)(=O)[O-])O.[Na+]

Specifications

102029-60-7
MFCD00067500
UVGYCHPTWHWEPZ-UHFFFAOYSA-M
sodium;2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
44119996
Odorless
Colorless
AMPSO buffer soln.
C7H16NNaO5S
0.2M buffer soln.
CC(C)(CO)NCC(CS(=O)(=O)[O-])O.[Na+]
249.257
Liquid
8.5
100mL
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C