Invitrogen™ AMPK (A2/B2/G2) Recombinant Human Protein DFS Item
AMP-activated protein kinase (AMPK) exhibits a key role as a master regulator of cellular energy homeostasis. AMPK exists as a heterotrimeric complex composed of a catalytic α subunit and regulatory β and γ subunits. Binding of AMP to the γ subunit allosterically activates the complex.
$612.00 - $9,380.00
Specifications
Host Species Human
Protein Tag His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6250
View Documents
Invitrogen™
LSPV6250
10μg Each for $612.00
PV6251
View Documents
Invitrogen™
LSPV6251
100μg Each for $2,095.00
PV6252
View Documents
Invitrogen™
LSPV6252
1mg Each for $9,380.00
Description
Description
AMPK is activated in response to stresses that deplete cellular ATP (low glucose, hypoxia and ischemia) and via signaling pathways in response to adiponectin, leptin and CAMKK β. Recombinant full-length human AMPK (combination of A2/B2/G2 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags.
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
His Tag
Yes