Please login to your online account to display your discounted pricing

Alfa Aesar™ Amphotericin B, 250 μg/mL in distilled water, sterile-filtered

$98.84 - $98.84

Chemical Identifiers

Chemical Name or Material Amphotericin B
CAS 1397-89-3
Name Note 250μg/mL in distilled water, sterile-filtered
MDL Number MFCD00877763
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67049AD

  SDS
alfa aesar™
J67049AD
50mL Each for $98.84
Description & Specifications

Specifications

Chemical Name or Material Amphotericin B
CAS 1397-89-3
Physical Form Liquid
MDL Number MFCD00877763
Name Note 250μg/mL in distilled water, sterile-filtered
Storage Note 1 -30° to -10°C
RTECS Number BU2625000
Quantity 50mL