Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Amphiregulin Ab-2, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in western blotting and Immunoprecipitation experiments with Thermo Scientific™ Amphiregulin Ab-2, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB258P1ABX

 View more versions of this product

Catalog No. RB258P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Amphiregulin Ab-2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic 19-mer peptide, corresponding to aa 26-44 from the secreted amphiregulin of human origin
Isotype IgG
Quantity 500μL
Regulatory Status RUO
Research Discipline EGFR Family
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Human
Target Species Murine

Obtain accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Amphiregulin Ab-2, Rabbit Polyclonal Antibody.

 

Amphiregulin (AR) binds to EGF-Receptor (EGFR) with lower affinity than EGF. The mature secreted form AR is an 84-amino acid residue glycosylated polypeptide growth regulator, which is generated by proteolytic processing of a 252-amino acid transmembrane precursor. Seven different polypeptide ligands, which derive from distinct genes, are capable of binding to the extracellular domain of EGFR. These ligands include EGF, TGFα, AR, HB-EGF, cripto-1, epiregulin, and βcellulin. All of these growth factors contain a characteristic EGF-like domain which is defined by 6 evenly spaced cysteine residues that generate 3 loops through the formation of disulfide bonds. AR protein has been localized by immunohistochemistry to the epithelium of the colon, stomach, pancreas, breast, and placenta. AR is reportedly overexpressed in human cancers of breast, colon, stomach, and pancreas.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 26-44

Immunogen: A synthetic 19-mer peptide, corresponding to aa 26-44 from the secreted amphiregulin of human origin

Molecular Weight: 18.5-22.5kDa

Positive Control: HT29, LS174T cells

Cellular Localization: Cytoplasmic and/or nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO