missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ AMPD, 0.2M buffer soln., pH 8.9

$77.57 - $121.51

Chemical Identifiers

CAS 115-69-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00004678
InChI Key UXFQFBNBSPQBJW-UHFFFAOYSA-N
PubChem CID 1531
ChEBI CHEBI:991
IUPAC Name 2-amino-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61377AE View Documents Alfa Aesar™
J61377AE
100mL Each for $77.57
 
AAJ61377AK View Documents Alfa Aesar™
J61377AK
250mL Each for $121.51
 
Specifications

Chemical Identifiers

115-69-5
105.137
UXFQFBNBSPQBJW-UHFFFAOYSA-N
CHEBI:991
CC(CO)(CO)N
C4H11NO2
MFCD00004678
1531
2-amino-2-methylpropane-1,3-diol

Specifications

115-69-5
MFCD00004678
UXFQFBNBSPQBJW-UHFFFAOYSA-N
2-amino-2-methylpropane-1,3-diol
1531
Liquid
8.9
100mL
C4H11NO2
0.2M buffer soln.
CC(CO)(CO)N
105.137
CHEBI:991
Odorless
Colorless
AMPD buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C