missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ AMP, 0.2M buffer soln., pH 9.0

$77.57 - $121.51

Chemical Identifiers

CAS 124-68-5
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008051
InChI Key CBTVGIZVANVGBH-UHFFFAOYSA-N
Synonym 2-Amino-2-methyl-1-propanol
PubChem CID 11807
IUPAC Name 2-amino-2-methylpropan-1-ol
SMILES CC(C)(CO)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62043AE View Documents Alfa Aesar™
J62043AE
100mL Each for $77.57
 
AAJ62043AK View Documents Alfa Aesar™
J62043AK
250mL Each for $121.51
 
Specifications

Chemical Identifiers

124-68-5
89.138
CBTVGIZVANVGBH-UHFFFAOYSA-N
11807
CC(C)(CO)N
C4H11NO
MFCD00008051
2-Amino-2-methyl-1-propanol
2-amino-2-methylpropan-1-ol

Specifications

124-68-5
MFCD00008051
2-Amino-2-methyl-1-propanol
CC(C)(CO)N
89.138
Liquid
9.0
100mL
C4H11NO
0.2M buffer soln.
CBTVGIZVANVGBH-UHFFFAOYSA-N
2-amino-2-methylpropan-1-ol
11807
Odorless
Colorless
AMP buffer soln.
Safety and Handling

Safety and Handling

RTECSNumber : UA5950000

TSCA : TSCA

Recommended Storage : 2° to 8°C