Learn More
Amoxicillin sodium salt, 89% min., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6667514
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Amoxicillin Sodium Salt | |
Powder | |
130°C | |
MFCD08063910 | |
BYHDFCISJXIVBV-ZONSQSGVSA-M | |
sodium;(2S,5R,6S)-6-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
131632470 | |
≥89% | |
25 g | |
743°C |
Amoxicillin sodium salt | |
34642-77-8 | |
C16H18N3NaO5S | |
Soluble in water. | |
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)[O-])C.[Na+] | |
387.386 | |
387.39 | |
White | |
>110°C (230°F) |
Chemical Identifiers
34642-77-8 | |
387.386 | |
BYHDFCISJXIVBV-ZONSQSGVSA-M | |
sodium;(2S,5R,6S)-6-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
C16H18N3NaO5S | |
MFCD08063910 | |
131632470 | |
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)[O-])C.[Na+] |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 252-124-1
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only