Please login to your online account to display your discounted pricing

AMN 082 dihydrochloride, Tocris

The first selective mGlu7 agonist

$142.08 - $598.33

Chemical Identifiers

Chemical Name or Material AMN 082 dihydrochloride
Assay Percent Range >99%
Molecular Formula C28H28N2·2HCl
Formula Weight 465.45
Synonym N, N'-Bis(diphenylmethyl)-1, 2-ethanediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

238550

 
r & d systems
RDS238550
50mg Each for $598.33

238510

 
r & d systems
RDS238510
10mg Each for $142.08
Description & Specifications

Specifications

Formula Weight 465.45
Quantity 50mg
Chemical Name or Material AMN 082 dihydrochloride
CAS 97075-46-2
Assay Percent Range >99%
Molecular Formula C28H28N2·2HCl
Synonym N, N'-Bis(diphenylmethyl)-1, 2-ethanediamine dihydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C