missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium vanadium oxide, 99% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01109130

 View more versions of this product

Catalog No. AA1109130



Description

Description

Ammonium vanadium oxide is used as a catalyst in organic synthesis, and as an analytical reagent in laboratories. It is employed in the preparation of vanadate. It acts as a sensor for the determination of N-acetyltransferase activity in humans. Further, it finds use in dyes, varnishes, indelible inks, drier for paints, and in photography. With copper carbonate, it has been shown to enhance the catalytic activities. It is an environmental-friendly catalyst to prepare alfa-hydroxyphosphonate derivatives in high yield through the reaction of aryl/heteroaryl aldehydes with triethylphosphonate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium vanadium oxide
7803-55-6
Powder
99% (Metal basis)
MFCD00011430
Slightly soluble in water. Soluble in dilute ammonia and diethanolamine. Insoluble in ether,and alcohol.
N.O[V](=O)=O
116.98
116.98
2.33 g/cm3
200°C (decomposition)
White
250 g
H4NO3V
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
YBVKNHXQSRDWAA-UHFFFAOYSA-M
vanadiumoylol amine
516859
Off-white powder

Chemical Identifiers

7803-55-6
116.98
YBVKNHXQSRDWAA-UHFFFAOYSA-M
516859
H4NO3V
MFCD00011430
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
N.O[V](=O)=O
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c

H301-H319-H332-H361d-H372

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 232-261-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only