missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium tungsten oxide pentahydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Chemical Identifiers

CAS 1311-93-9
Physical Form Crystalline Powder
Assay Percent Range 99.999% (Metal basis)
Molecular Formula (NH4)10W12O41·5H2O
Formula Weight 3132.64
MDL Number MFCD00015956
Chemical Name or Material Ammonium tungsten oxide pentahydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1089914 SDS Alfa Aesar
1089914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1089922 SDS Alfa Aesar
1089922
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1311-93-9
99.999% (Metal basis)
3132.64
Ammonium tungsten oxide pentahydrate
Crystalline Powder
(NH4)10W12O41·5H2O
MFCD00015956

Specifications

Crystalline Powder
25g
Odorless
(NH4)10W12O41·5H2O
1311-93-9
MFCD00015956
3132.64
White
2.3g/cm3
99.999% (Metal basis)
White crystalline
Ammonium tungsten oxide pentahydrate

Safety and Handling

EINECSNumber : 234-364-9