Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tungsten oxide hydrate

Chemical Identifiers

Chemical Name or Material Ammonium tungsten oxide hydrate
CAS 11120-25-5
Linear Formula (NH4)10W12O41·xH2O
Formula Weight 3042.56 (Anhydrous)
MDL Number MFCD00015956
Synonym Ammonium paratungstate; Ammonium tungstate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2264014

  SDS
alfa aesar™
2264014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 3042.56 (Anhydrous)
Linear Formula (NH4)10W12O41·xH2O
Synonym Ammonium paratungstate; Ammonium tungstate
CAS 11120-25-5
Physical Form Powder
Density 2.3g/mL
Quantity 25g
Solubility Information Soluble in water; Insoluble in alcohol
TSCA TSCA
Color White
Odor Odorless
Chemical Name or Material Ammonium tungsten oxide hydrate
MDL Number MFCD00015956