missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tungsten oxide, 99.99+% (metals basis)

Manufacturer:  Alfa Aesar™ 4458809

 View more versions of this product

Catalog No. AA4458809



Specifications

Specifications

Ammonium tungsten oxide
(metals basis)
MFCD00015956
Soluble in warm water
N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O
71306883
Powder
10g
11140-77-5
H40N10O41W12
ammonium para tungstate hydrate
LMAJGSPZJYEENT-UHFFFAOYSA-D
3042.429
283.91
≥99.99%

Chemical Identifiers

11140-77-5
3042.429
LMAJGSPZJYEENT-UHFFFAOYSA-D
71306883
H40N10O41W12
MFCD00015956
ammonium para tungstate hydrate
N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents