missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium tungsten oxide, 99.99+% (metals basis), Thermo Scientific Chemicals
$173.90 - $3,207.79
Chemical Identifiers
CAS | 11140-77-5 |
---|---|
Molecular Formula | H40N10O41W12 |
Molecular Weight (g/mol) | 3042.429 |
MDL Number | MFCD00015956 |
InChI Key | LMAJGSPZJYEENT-UHFFFAOYSA-D |
Synonym | ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine |
PubChem CID | 71306883 |
SMILES | N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O |
Description
Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11140-77-5 | |
3042.429 | |
LMAJGSPZJYEENT-UHFFFAOYSA-D | |
71306883 |
H40N10O41W12 | |
MFCD00015956 | |
ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine | |
N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O |
Specifications
11140-77-5 | |
10 g | |
MFCD00015956 | |
Soluble in warm water | |
N.N.N.N.N.N.N.N.N.N.O[W]12(=O)O[W]3(=O)(O[W](=O)(O3)(O[W]4(=O)(O[W](=O)(O4)(O[W]5(=O)(O[W]6(=O)(O5)O[W]7(=O)(O6)O[W](=O)(O7)(O[W]8(=O)(O[W](=O)(O8)(O[W](=O)(O1)(O2)O)O)O)O)O)O)O)O)O | |
71306883 | |
≥99.99% | |
(metals basis) |
Powder | |
H40N10O41W12 | |
ammonium para tungstate hydrate, ammonium tungstate trace metals basis, ammonium paratungstate trace metals basis, dodecaoxooctacyclo 21.1.1.1 1 , 3 .1 3 ,?.1?,?.1 1 1 , 1 3 .1 1 ?, 1 ?.1 1 ?, 2 1 dodecatungstoxane-5,7,9,11,13,15,17,19,21,23-decol decaamine | |
LMAJGSPZJYEENT-UHFFFAOYSA-D | |
3042.429 | |
283.91 | |
Ammonium tungsten oxide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only