missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium trifluoromethanesulfonate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC388730500

 View more versions of this product

Catalog No. AC388730500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
50g
CH4F3NO3S
Ammonium trifluoromethanesulfonate
99%
Glass bottle
98.5% min.
167.11
White
Authentic
38542-94-8
MFCD00075332
Ammonium triflate

Chemical Identifiers

99%
167.11
Ammonium triflate
CH4F3NO3S
MFCD00075332
Ammonium trifluoromethanesulfonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.