Please login to your online account to display your discounted pricing

Ammonium titanyl oxalate monohydrate, 98%, pure, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC212731000

 View more versions of this product

Catalog No. AC212731000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥97.5% (calculated on oxalate)
Formula Weight 294.02
Quantity 100g
Color White to light beige
Purity Grade Pure
Chemical Name or Material Ammonium titanyl oxalate monohydrate
Linear Formula (NH4)2TiO(C2O4)2·H2O
MDL Number MFCD00149620
Synonym Ammonium bis(oxalato)oxotitanate(IV)
Health Hazard 2 GHS P Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Packaging Plastic bottle
Assay Percent Range 98%
Infrared Spectrum Authentic
Molecular Formula C4H8N2O9Ti·H2O
CAS 10580-03-7
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material Ammonium titanyl oxalate monohydrate
CAS 10580-03-7
Purity Grade Pure
Assay ≥97.5% (calculated on oxalate)
Assay Percent Range 98%
Molecular Formula C4H8N2O9Ti·H2O
Linear Formula (NH4)2TiO(C2O4)2·H2O
Formula Weight 294.02
MDL Number MFCD00149620
Synonym Ammonium bis(oxalato)oxotitanate(IV)
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

SDS & Safety Documents