missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiosulfate, 98%+, MilliporeSigma™

Manufacturer:  Emd Millipore Corporation 1012089025

 View more versions of this product

Catalog No. M1012089025



Specifications

Specifications

Ammonium Thiosulfate
98%+
White solid
148.19
Odorless
25kg
7783-18-8
(NH4)2S2O3
acmc-1bhnz, ammonium thiosulphate, ammoniumthiosulfate, diammonium hypo, diammonium thiosulphate, dsstox_cid_9703, dsstox_gsid_29703, dsstox_rid_78814, thiosulfuric acid h2s2o3 , ammonium salt 1:2, thiosulfuric acid h2s2o3 , diammonium salt
Solid
Fibre carton

Chemical Identifiers

7783-18-8
(NH4)2S2O3
acmc-1bhnz, ammonium thiosulphate, ammoniumthiosulfate, diammonium hypo, diammonium thiosulphate, dsstox_cid_9703, dsstox_gsid_29703, dsstox_rid_78814, thiosulfuric acid h2s2o3 , ammonium salt 1:2, thiosulfuric acid h2s2o3 , diammonium salt
Documents