missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiosulfate, 98%+, MilliporeSigma™

Manufacturer:  MilliporeSigma 1012081000

 View more versions of this product

Catalog No. M1012081000



Specifications

Specifications

Ammonium Thiosulfate
98%+
White solid
148.19
Odorless
1kg
7783-18-8
(NH4)2S2O3
acmc-1bhnz
Solid
Plastic bottle

Chemical Identifiers

7783-18-8
(NH4)2S2O3
acmc-1bhnz
Documents