Please login to your online account to display your discounted pricing

Ammonium Thiosulfate, 98%+, EMD Millipore™

$205.66 - $2,469.40

Chemical Identifiers

Chemical Name or Material Ammonium Thiosulfate
CAS 7783-18-8
Molecular Formula (NH4)2S2O3
Formula Weight 148.19
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1012081000

 
emd millipore corporation
1012081000
Each for $205.66

M1012089025

 
emd millipore corporation
1012089025
Each for $2,469.40
Description & Specifications

Specifications

Formula Weight 148.19
Molecular Formula (NH4)2S2O3
CAS 7783-18-8
Appearance White solid
Chemical Name or Material Ammonium Thiosulfate
Packaging Plastic bottle
Assay Percent Range 98%+
Odor Odorless
Physical Form Solid