Please login to your online account to display your discounted pricing

Ammonium thioglycolate, 70 wt.% solution in water, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384682500

 View more versions of this product

Catalog No. AC384682500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 109.15
Quantity 250mL
Color Clear colorless
Assay 68.0 to 72.0%
pH 5.8 to 6.4
Density 1.22g/mL
Iron (Fe) 0.2ppm max.
Chemical Name or Material Ammonium thioglycolate
Name Note 70 wt.% solution in water
Molecular Formula C2H7NO2S
Synonym Ammonium mercaptoacetate, 70 wt.% solution in water
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May be corrosive to metals.
CAS 7732-18-5
CAS 5421-46-5
Identification ammonium: positive, mercaptane: positive
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00137451

Specifications

Chemical Name or Material Ammonium thioglycolate
Name Note 70 wt.% solution in water
CAS 5421-46-5
CAS 7732-18-5
Assay 68.0 to 72.0%
Molecular Formula C2H7NO2S
Formula Weight 109.15
MDL Number MFCD00137451
Synonym Ammonium mercaptoacetate, 70 wt.% solution in water
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May be corrosive to metals.

SDS & Safety Documents