Please login to your online account to display your discounted pricing

Ammonium thioglycolate, 70 wt.% solution in water, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384680010

 View more versions of this product

Catalog No. AC384680010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 109.15
Quantity 1L
Color Clear colorless
Assay 68.0 to 72.0%
pH 5.8 to 6.4
Density 1.22g/mL
Iron (Fe) 0.2ppm max.
CAS 7732-18-5
Chemical Name or Material Ammonium thioglycolate
Name Note 70 wt.% solution in water
Molecular Formula C2H7NO2S
MDL Number MFCD00137451
CAS 5421-46-5
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May be corrosive to metals.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym Ammonium mercaptoacetate, 70 wt.% solution in water
Identification ammonium: positive, mercaptane: positive
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Ammonium thioglycolate
Name Note 70 wt.% solution in water
CAS 5421-46-5
CAS 7732-18-5
Assay 68.0 to 72.0%
Molecular Formula C2H7NO2S
Formula Weight 109.15
MDL Number MFCD00137451
Synonym Ammonium mercaptoacetate, 70 wt.% solution in water
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May be corrosive to metals.

SDS & Safety Documents