Please login to your online account to display your discounted pricing

Ammonium Thiocyanate, Certified, 30.0% (w/v) ±0.5% (w/v), LabChem™

$148.40 - $148.40

Chemical Identifiers

Chemical Name or Material Ammonium Thiocyanate
CAS 7732-18-5
CAS Max % 70
CAS 1762-95-4
CAS Max % 30
Purity Grade Certified Grade
Linear Formula NH4SCN
Formula Weight 76.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC113451

 
lab chem
LC113451
500mL Each for $148.40
Description & Specifications

Specifications

Formula Weight 76.12
Purity Grade Certified Grade
Assay 30.0% ± 0.5% (w/v)
Chemical Name or Material Ammonium Thiocyanate
Solubility Soluble in water
CAS Max % 70
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Color Clear, colorless
CAS 1762-95-4
Health Hazard 1 Warning
Decomposition Information Gaseous Ammonia; Sulfur compounds; Nitrogen oxides; Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Storage Information Room Temperature
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Physical Form Liquid
Linear Formula NH4SCN
Identification Passes Test
Quantity 500mL
CAS Max % 30
Traceability to NIST Traceable to NIST
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.