missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiocyanate, Certified, 0.0250N ±0.0004N (0.025M), LabChem™

$100.57 - $100.57

Chemical Identifiers

CAS 7732-18-5,1762-95-4
CAS Max % 99.81,0.19
Linear Formula NH4SCN
Formula Weight 76.12
Chemical Name or Material Ammonium Thiocyanate
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC113501 Lab Chem
LC113501
500mL Each for $100.57
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1762-95-4
NH4SCN
Ammonium Thiocyanate
99.81,0.19
76.12
Certified Grade

Specifications

76.12
500mL
Passes Test
Gaseous Ammonia; Sulfur compounds; Nitrogen oxides; Carbon monoxide; Carbon dioxide
Clear, colorless
Room Temperature
NH4SCN
Certified Grade
1762-95-4,7732-18-5
99.81,0.19
Soluble in water
0.0250N ±0.0004N (0.025M)
Poly Bottle
Liquid
Ammonium Thiocyanate

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.