missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiocyanate, ACS Grade, ≥ 97.5%, LabChem™

$178.26 - $178.26

Chemical Identifiers

CAS 1762-95-4
CAS Max % 100
Linear Formula NH4SCN
Formula Weight 76.12
Chemical Name or Material Ammonium Thiocyanate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC113401 Lab Chem
LC113401
500g Each for $178.26
Specifications Description & Specifications

Chemical Identifiers

1762-95-4
NH4SCN
Ammonium Thiocyanate
100
76.12
ACS Grade

Specifications

1762-95-4
Crystals
76.12
Passes Test
Soluble in water
Room Temperature
White
500g
Ammonium Thiocyanate
ACS Grade
100
NH4SCN
1.3g/cm3
Poly Bottle
1.3g/cm3
Nitrogen oxides; Carbon dioxide; Carbon monoxide; Sulfur compounds; Gaseous Ammonia
150°C
190°C
≥97.5%

Safety and Handling

GHS H Statement
Harmful if swallowed, in contact with skin or if inhaled.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

EINECSNumber : 217-175-6