Learn More
Ammonium thiocyanate, 99+%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 206500010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ammonium thiocyanate | |
1kg | |
CH4N2S | |
MFCD00011428 | |
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
SOIFLUNRINLCBN-UHFFFAOYSA-N | |
azanium;thiocyanate | |
15666 | |
99+% | |
Plastic bottle | |
1.3000g/mL |
1762-95-4 | |
99% min. (on dried basis) (Argentometry) | |
NH4SCN | |
15,558 | |
Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol,acetone and ammonia,practically insoluble in chcl3 and ethyl acetate | |
C(#N)[S-].[NH4+] | |
76.12 | |
76.12 | |
Extra Pure | |
1.3 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF
GHS Signal Word: Danger
EINECSNumber : 217-175-6
RUO â Research Use Only