missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Thiocyanate, 0.100 Normal, Ricca Chemical

$37.21 - $177.06

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium isothiocyanate
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R0695000500 View Documents Ricca Chemical Company
RIR0695000500A
500mL Poly natural Each for $37.21
 
695-1 SDS View Documents Ricca Chemical Company
RI6951
4L Poly natural Each for $177.06
 
R06950004F View Documents Ricca Chemical Company
RIR06950004F
4L Cubitainer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R069500020F View Documents Ricca Chemical Company
RIR069500020F
20L Cubitainer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1762-95-4
76.117
ammonium isothiocyanate
azanium;thiocyanate
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
15666
C(#N)[S-].[NH4+]

Specifications

1762-95-4,7732-18-5
100.0,0.77
ammonium isothiocyanate
C(#N)[S-].[NH4+]
76.117
Liquid
0.100Normal
500mL
97.0,0.74
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
azanium;thiocyanate
15666
Poly natural
Colorless
Ammonium Thiocyanate