Please login to your online account to display your discounted pricing

Ammonium tetrathiotungstate, Yellow chunks/powder, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Ammonium tetrathiotungstate
CAS 13862-78-7
Physical Form Chunks and Powder
Molecular Formula (NH4)2WS4
Formula Weight 348.07
MDL Number MFCD00061410
Synonym Ammonium thiotungstate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2237304

  SDS
alfa aesar
2237304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2237309

  SDS
alfa aesar
2237309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Chunks and Powder
Formula Weight 348.07
Quantity 2g
Color Yellow
Chemical Name or Material Ammonium tetrathiotungstate
Odor Like rotten eggs
Molecular Formula (NH4)2WS4
MDL Number MFCD00061410
CAS 13862-78-7
Synonym Ammonium thiotungstate
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.