missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium tetrathiomolybdate, 99.95%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389530100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Ammonium tetrathiomolybdate | |
White | |
Crystals or Powder | |
99.95% | |
(NH4)2MoS4 | |
15,557 | |
ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
diazanium;bis(sulfanylidene)molybdenum;sulfanide | |
15251598 | |
99.95% | |
Glass bottle |
300.0°C | |
15060-55-6 | |
10 g | |
H10MoN2S4 | |
MFCD00136013 | |
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S | |
262.28 | |
260.28 | |
Trace Metal Basis |
Chemical Identifiers
15060-55-6 | |
262.28 | |
ZKKLPDLKUGTPME-UHFFFAOYSA-N | |
15251598 |
H10MoN2S4 | |
MFCD00136013 | |
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4 | |
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only