missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium tetrathiomolybdate, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4349306

 View more versions of this product

Catalog No. AA4349306



Specifications

Specifications

Ammonium tetrathiomolybdate
99.95% (metals basis)
MFCD00136013
ZKKLPDLKUGTPME-UHFFFAOYSA-N
diazanium;bis(sulfanylidene)molybdenum;sulfanide
15251598
Crystalline
15060-55-6
H10MoN2S4
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
262.284
260.27
5g

Chemical Identifiers

15060-55-6
262.284
ZKKLPDLKUGTPME-UHFFFAOYSA-N
15251598
[NH4+].[NH4+].[SH-].[SH-].S=[Mo]=S
H10MoN2S4
MFCD00136013
ammonium tetrathiomolybdate, molybdate 2-, tetrathioxo-, ammonium 1:2 , t-4, tiomolibdate diammonium, unii-4v6i63lw1e, ammonium thiomolybdate, molybdate 2-, tetrathioxo-, diammonium, t-4, ammonium tetrathiomolybdate vi, molybdate 2-, tetrathioxo-, diammonium, beta-4
diazanium;bis(sulfanylidene)molybdenum;sulfanide
Documents