missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetraphenylborate, 99%

Manufacturer:  Alfa Aesar™ H5570006

 View more versions of this product

Catalog No. AAH5570006



Specifications

Specifications

Ammonium tetraphenylborate
5g
C24H24BN
MFCD00134523
ammonium tetraphenylborate
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[NH4+]
337.273
337.27
220°C (decomposition)
14637-34-4
NH4B(C6H5)4
3799683
ZWFYDLYRTYMBIL-UHFFFAOYSA-O
azanium;tetraphenylboranuide
14496924
99%

Chemical Identifiers

14637-34-4
337.273
ZWFYDLYRTYMBIL-UHFFFAOYSA-O
14496924
[B-](C1=CC=CC=C1)(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[NH4+]
C24H24BN
MFCD00134523
ammonium tetraphenylborate
azanium;tetraphenylboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents