missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetrafluoroborate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4405522

 View more versions of this product

Catalog No. AA4405522



Specifications

Specifications

Ammonium tetrafluoroborate
(metals basis)
MFCD06245785
ammonium borofluoride
PDTKOBRZPAIMRD-UHFFFAOYSA-O
azanium;tetrafluoroborate
9964072
Crystalline
1.871 g/mL
Hygroscopic
220°C (sublimation)
13826-83-0
BF4H4N
UN3260
Soluble in water at 16°C 25g/L.
[B-](F)(F)(F)F.[NH4+]
104.843
104.84
99.999%
Ammonia-like
White
100g

Chemical Identifiers

13826-83-0
104.843
PDTKOBRZPAIMRD-UHFFFAOYSA-O
9964072
[B-](F)(F)(F)F.[NH4+]
BF4H4N
MFCD06245785
ammonium borofluoride
azanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H318-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 237-531-4

RTECSNumber : BQ6100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Documents