missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetrafluoroborate, 97%

Manufacturer:  Alfa Aesar™ 11488A3

 View more versions of this product

Catalog No. AA11488A3



Specifications

Specifications

Ammonium tetrafluoroborate
BF4H4N
UN3260
PDTKOBRZPAIMRD-UHFFFAOYSA-O
azanium;tetrafluoroborate
9964072
Powder
1.871g/mL
Hygroscopic
220°C (sublimation)
13826-83-0
MFCD06245785
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
[B-](F)(F)(F)F.[NH4+]
104.843
104.84
97%
Ammonia-like
White
2kg

Chemical Identifiers

13826-83-0
104.843
PDTKOBRZPAIMRD-UHFFFAOYSA-O
9964072
[B-](F)(F)(F)F.[NH4+]
BF4H4N
MFCD06245785
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p
azanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : BQ6100000

TSCA : TSCA

Documents