missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium tetrachloroplatinate(II), 99.998%, (trace metal basis), ACROS Organics™

Chemical Identifiers

CAS 13820-41-2
Assay Percent Range 99.998%
Molecular Formula H8Cl4N2Pt
Linear Formula (NH4)2PtCl4
Formula Weight 372.97
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Assay 51.5 to 53.1% (Pt)
Grade Trace Metal Basis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC193750010 SDS Thermo Fisher Scientific
AC193750010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For ultra trace analysis
Description & Specifications

Chemical Identifiers

13820-41-2
H8Cl4N2Pt
372.97
51.5 to 53.1% (Pt)
99.998%
(NH4)2PtCl4
Ammonium tetrachloroplatinate(II)
Trace Metal Basis

Specifications

Pink
372.97
2.9300g/mL
20ppm max.
13820-41-2
Glass bottle
51.5 to 53.1% (Pt)
Crystals
1g
(NH4)2PtCl4
H8Cl4N2Pt
99.998%
Ammonium tetrachloroplatinate(II)
Trace Metal Basis

Safety and Handling

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger