missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetrachloroplatinate(II), 99.9% (metals basis), Pt 51% min

Manufacturer:  Alfa Aesar™ 1104603

 View more versions of this product

Catalog No. AA1104603



Specifications

Specifications

Ammonium tetrachloroplatinate(II)
(metals basis)
MFCD00010885
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
Crystalline
Pt 51% min.
Odorless
145°C (decomposition)
13820-41-2
(NH4)2PtCl4
UN3288
372.98
99.9%
2.936g/mL
Red
1g

Chemical Identifiers

13820-41-2
MFCD00010885
ammonium tetrachloroplatinate ii, diammonium tetrachloroplatinate, ammonium platinous chloride, diammonium tetrachloroplatinate 2-, ammonium tetrachloroplatinate, bis ammonium tetrachloroplatinate 2-, ammonium chloroplatinate ii, unii-z5ny20a9s2, h8cl4n2pt, epitope id:151521
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H318-H290-H315-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : TP1840000

TSCA : TSCA

Documents