Please login to your online account to display your discounted pricing

Ammonium tetrachloroplatinate(II), 51 to 54% Pt, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195390050

 View more versions of this product

Catalog No. AC195390050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 372.97
Quantity 5g
Color Light brown orange to red
Merck Index 15, 546
EINECS Number 237-499-1
TSCA TSCA
Assay 51 to 54% (Pt)
Density 2.93g/mL
RTECS Number TP1840000
CAS 13820-41-2
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Ammonium tetrachloroplatinate(II)
Name Note 51 to 54% Pt
Packaging Glass bottle
Molecular Formula H8Cl4N2Pt
Linear Formula (NH4)2PtCl4
MDL Number MFCD00010885
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Ammonium tetrachloroplatinate(II)
Name Note 51 to 54% Pt
CAS 13820-41-2
Assay 51 to 54% (Pt)
Molecular Formula H8Cl4N2Pt
Linear Formula (NH4)2PtCl4
Formula Weight 372.97
MDL Number MFCD00010885
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.

SDS & Safety Documents