Please login to your online account to display your discounted pricing

Ammonium tetrachloroplatinate(II), 51 to 54% Pt, ACROS Organics™

$173.67 - $173.67

Chemical Identifiers

Chemical Name or Material Ammonium tetrachloroplatinate(II)
Name Note 51 to 54% Pt
CAS 13820-41-2
Assay 51 to 54% (Pt)
Molecular Formula H8Cl4N2Pt
Linear Formula (NH4)2PtCl4
Formula Weight 372.97
MDL Number MFCD00010885
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195390010

  SDS
thermo fisher scientific
195390010
Glass bottle 1g Each for $173.67

AC195390050

  SDS
thermo fisher scientific
195390050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 372.97
Physical Form Crystalline Powder
Quantity 1g
Color Light brown orange to red
Assay 51 to 54% (Pt)
RTECS Number TP1840000
TSCA TSCA
EINECS Number 237-499-1
Density 2.93g/mL
Merck Index 15, 546
Packaging Glass bottle
Linear Formula (NH4)2PtCl4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.
Chemical Name or Material Ammonium tetrachloroplatinate(II)
MDL Number MFCD00010885
Molecular Formula H8Cl4N2Pt
CAS 13820-41-2
Name Note 51 to 54% Pt
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic if swallowed.