missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%

Manufacturer:  Alfa Aesar™ 1082403

 View more versions of this product

Catalog No. AA1082403



Specifications

Specifications

Ammonium tetrachloropalladate(II)
(metals basis)
(NH4)2PdCl4
ammonium chloropalladite
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl
284.298
CHEBI:60149
Powder
Premion™
2.17g/mL
Brown to Green
13820-40-1
Cl4H8N2Pd
MFCD00010884
IRKVTUVFHGUMMN-UHFFFAOYSA-L
diazanium;tetrachloropalladium(2-)
61682
284.31
99.998%
Pd 37%
Hygroscopic
1g

Chemical Identifiers

13820-40-1
284.298
IRKVTUVFHGUMMN-UHFFFAOYSA-L
61682
diazanium;tetrachloropalladium(2-)
Cl4H8N2Pd
MFCD00010884
ammonium chloropalladite
CHEBI:60149
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P310-P362-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
mmediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BP5400000

TSCA : TSCA

Documents