Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium tetrachloropalladate(II), Premion™, 99.998% (metals basis), Pd 37%

$387.89 - $387.89

Chemical Identifiers

Chemical Name or Material Ammonium tetrachloropalladate(II)
CAS 13820-40-1
Purity Grade Premion™
Linear Formula (NH4)2PdCl4
Formula Weight 284.31
MDL Number MFCD00010884
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1082403

 
alfa aesar™
1082403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1082406

 
alfa aesar™
1082406
5g Each for $387.89

AA1082414

 
alfa aesar™
1082414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.31
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310-P362-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
mmediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ammonium tetrachloropalladate(II)
TSCA TSCA
Purity Grade Premion™
Physical Form Powder
Assay 99.998%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula (NH4)2PdCl4
MDL Number MFCD00010884
CAS 13820-40-1
Color Dark green to brown
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Concentration Pd 37%
Density 2.17g/mL
RTECS Number BP5400000
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310-P362-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
mmediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H315
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.