missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium tartrate, 98.8%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0219504280
Specifications
Ammonium tartrate | |
0.988 | |
ammonium d-tartrate, ammonium l-+-tartrate, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, diammonium 2r,3r-2,3-dihydroxysuccinate, diammonium l-+-tartrate, diammonium l-tartrate, l-tartaric acid, ammonium salt, r-2,3-dihydroxybutanedioic acid diammonium salt, unii-ji8ud88354 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+] | |
184.148 | |
CHEBI:63075 | |
Crystalline Powder | |
100g |
3164-29-2 | |
C4H12N2O6 | |
NGPGDYLVALNKEG-OLXYHTOASA-N | |
diazanium;(2R,3R)-2,3-dihydroxybutanedioate | |
13652278 | |
184.1 | |
White |
Chemical Identifiers
3164-29-2 | |
184.148 | |
ammonium d-tartrate, ammonium l-+-tartrate, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, diammonium 2r,3r-2,3-dihydroxysuccinate, diammonium l-+-tartrate, diammonium l-tartrate, l-tartaric acid, ammonium salt, r-2,3-dihydroxybutanedioic acid diammonium salt, unii-ji8ud88354 | |
CHEBI:63075 | |
C(C(C(=O)[O-])O)(C(=O)[O-])O.[NH4+].[NH4+] |
C4H12N2O6 | |
NGPGDYLVALNKEG-OLXYHTOASA-N | |
13652278 | |
diazanium;(2R,3R)-2,3-dihydroxybutanedioate |