missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium sulfite monohydrate, 92% min Alfa Aesar

Air Sensitive

Manufacturer:  Alfa Aesar 17996A1

 View more versions of this product

Catalog No. AA17996A1



Specifications

Specifications

Ammonium sulfite monohydrate
7783-11-1
H10N2O4S
White crystalline
[NH4+].[NH4+].O.[O-]S(=O)[O-]
134.15
134.15
1.41g/cm3
1kg
Monohydrate
92% min.
MFCD00066094
ASPXBYAQZVXSNS-UHFFFAOYSA-N
diazanium;sulfite;hydrate
21701053
Crystalline
White

Chemical Identifiers

7783-11-1
134.15
ASPXBYAQZVXSNS-UHFFFAOYSA-N
diazanium;sulfite;hydrate
H10N2O4S
MFCD00066094
21701053
[NH4+].[NH4+].O.[O-]S(=O)[O-]
Safety and Handling

Safety and Handling

H315-H319-H335

EINECSNumber : 233-484-9

SDS Documents