Please login to your online account to display your discounted pricing

Ammonium sulfite monohydrate, 92% min Alfa Aesar

Air Sensitive

$57.41 - $57.41

Chemical Identifiers

Chemical Name or Material Ammonium sulfite monohydrate
Name Note Monohydrate
CAS 7783-11-1
Physical Form Crystalline
Assay Percent Range 92% min.
Molecular Formula H8N2O3S·H2O
Formula Weight 134.15
MDL Number MFCD00066094
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA17996A1

  SDS
alfa aesar
17996A1
1kg Each for $57.41

AA17996A7

  SDS
alfa aesar
17996A7
5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.15
Quantity 1kg
Color White
Density 1.41g/cm3
Chemical Name or Material Ammonium sulfite monohydrate
EINECS Number 233-484-9
Name Note Monohydrate
Physical Form Crystalline
Assay Percent Range 92% min.
Molecular Formula H8N2O3S·H2O
Appearance White crystalline
MDL Number MFCD00066094
CAS 7783-11-1
Health Hazard 1 H315-H319-H335
Safety and Handling

H315-H319-H335