missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium sulfide, 20% solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 426080010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium sulfide | |
12135-76-1 | |
H8N2S | |
MFCD00010892 | |
15, 553 | |
UXXGWUQNRMPNKH-UHFFFAOYSA-N | |
diamine sulfane | |
68.14 | |
Glass bottle | |
1 |
20% solution in water | |
1 L | |
(NH4)2S | |
32°C | |
Solubility in water: miscible | |
N.N.S | |
68.14 | |
20.0 to 24.0% (Iodimetry) | |
Clear yellow solution | |
1.0000g/mL |
RUO – Research Use Only