missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium sulfide, 40-44% w/w aq. soln., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 040509AP

 View more versions of this product

Catalog No. AA40509AP



Description

Description

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth.It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium sulfide
12135-76-1
500 mL
MFCD00010892
UXXGWUQNRMPNKH-UHFFFAOYSA-N
diamine sulfane
68.14
Yellow to orange liquid
Yellow to Orange
Liquid
H8N2S
Miscible in water,alcohol and liquid ammonia.
N.N.S
68.14
Stench
546 hPa (410mm Hg) at 20°C

Chemical Identifiers

12135-76-1
68.14
UXXGWUQNRMPNKH-UHFFFAOYSA-N
N.N.S
H8N2S
MFCD00010892
diamine sulfane
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 235-223-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only