Please login to your online account to display your discounted pricing

Ammonium sulfate, Reagent grade, 99%, MP Biomedicals™


Quantity 500g

Manufacturer: mp biomedicals  10477290

 View more versions of this product

Catalog No. ICN10477290

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Reagent Grade
Physical Form Crystalline Powder
Formula Weight 132.1
Color White
Quantity 500g
Density 1.77g/cm3 at 20°C
Chemical Name or Material Ammonium sulfate
Assay Percent Range 0.99
Molecular Formula (NH4)2SO4
CAS 7783-20-2
Synonym Sulfuric acid diammonium salt

Specifications

Chemical Name or Material Ammonium sulfate
CAS 7783-20-2
Purity Grade Reagent Grade
Physical Form Crystalline Powder
Assay Percent Range 0.99
Molecular Formula (NH4)2SO4
Formula Weight 132.1
Synonym Sulfuric acid diammonium salt